Learn More
Sino Biological Influenza A H5N8 (A/duck/NY/191255-59/2002) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11717V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 527) of the influenza A H5N8 hemagglutinin (A/duck/NY/191255-59/2002 (H5N8)) (AAP72011.1), (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7368