Learn More
Sino Biological Influenza A H5N8 (A/duck/NY/191255-59/2002) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11717V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of the influenza A H5N8 hemagglutinin (A/duck/NY/191255-59/2002 (H5N8)) (AAP72011.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7369