Learn More
Sino Biological Influenza A H12N5 (A/green-winged teal/ALB/199/1991) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11718V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of the influenza A hemagglutinin (A/green-winged teal/ALB/199/1991 (H12N5)) (ABB88110.1), termed as HA1, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7374