Learn More
Sino Biological Influenza A H9N2 (A/Guinea fowl/Hong Kong/WF10/99) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11719V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 523) of the influenza A H9N2 Hemagglutinin (A/Guinea fowl/Hong Kong/WF10/99 (H9N2)) (AAO46082.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7380