Learn More
Sino Biological Influenza A H9N2 (A/Guinea fowl/Hong Kong/WF10/99) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11719V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 338) of the influenza hemagglutinin (A/Guinea fowl/Hong Kong/WF10/99 (H9N2)) (AAO46082.1), termed as HA1, was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7382