Learn More
Sino Biological Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11720V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Val 534) of the influenza A H15N8 hemagglutinin (A/duck/AUS/341/1983 (H15N8)) (ABB88132.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7386