Learn More
Sino Biological Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11720V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 349) of the influenza A hemagglutinin (A/duck/AUS/341/1983 (H15N8)) (ABB88132.1), termed as HA1, was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7388