Learn More
Sino Biological Influenza A H8N4 (A/pintail duck/Alberta/114/1979) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 11722V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/pintail duck/Alberta/114/1979(H8N4)) hemagglutinin (ABB87729.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7398