Learn More
Sino Biological Influenza A H6N1 (A/northern shoveler/California/HKWF115/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11723V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 529) of the influenza A H6N1 hemagglutinin (A/northern shoveler/California/ HKWF115/2007 (H6N1)) (ACE81692.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7406