Learn More
Sino Biological Influenza A H6N1 (A/northern shoveler/California/HKWF115/2007) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11723V08H1100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Arg 344) of the influenza A hemagglutinin (A/northern shoveler/California/HKWF115/2007 (H6N1)) (ACE81692.1), termed as HA1 was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-7407