Learn More
Sino Biological Recombinant Human LOK Protein (His Tag)
Supplier:  Sino Biological 11724H07E50
Encompass_Preferred
A DNA sequence encoding the human STK10 (NP_005981.3) (Arg 18-Glu 317) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7411