Learn More
Sino Biological Recombinant Human VNN2 Protein (His Tag)
Supplier:  Sino Biological 11728H08H50
Encompass_Preferred
A DNA sequence encoding the human VNN2 isoform 1 (NP_004656.2) without the propeptide (Met 1-Ser 492) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7421