Learn More
Sino Biological Recombinant Human CEACAM-8/CD66b Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11729H08H100
Encompass_Preferred
A DNA sequence encoding the human CEACAM8 (NP_001807.2) without the pro peptide (Met 1-Ser319) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7422