Learn More
Sino Biological Recombinant Human TEM7 Protein (His Tag)
Supplier:  Sino Biological 11736H08H50
Encompass_Preferred
A DNA sequence encoding the human PLXDC1 (AAH36059.1) extracellular domain (Met 1-Thr 426) was expressed, with a polyhistidine tag at the C-terminus
Catalog No. 50-161-7429