Learn More
Sino Biological Recombinant Human CaMKII beta/CAMK2B Protein (His & GST Tag)
Supplier:  Sino Biological 11738H20B50
Encompass_Preferred
A DNA sequence encoding the human CAMK2B isoform 2 (Q13554-3) (Met 1-Gln 503) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7431