Learn More
Sino Biological Recombinant Human PDE9A Protein (His Tag)
Supplier:  Sino Biological 11745H07E100
Encompass_Preferred
A DNA sequence encoding the human PDE9A (O76083-2) C-terminal segment (Pro 181-Lys 506) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7434