Learn More
Sino Biological Recombinant Human PDE1B Protein (His & GST Tag)
Supplier:  Sino Biological 11746H20B50
Encompass_Preferred
A DNA sequence encoding the human PDE1B long isoform (Q01064-1) (Met 1-Asp 536) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7439