Learn More
Sino Biological Recombinant Human SIRT1 Protein (His Tag)
Supplier:  Sino Biological 11748H07E100
Encompass_Preferred
A DNA sequence encoding the human SIRT1 (Met 193-Ser 747) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7444