Learn More
Sino Biological Recombinant Human Munc18-1/STXBP1 Protein (His & GST Tag)
Supplier:  Sino Biological 11751H20B100
Encompass_Preferred
A DNA sequence encoding the human STXBP1 isoform 1 (P61764-1) (Met 1-Ser 594) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7446