Learn More
Sino Biological Recombinant Human GRK2 Protein (His & GST Tag)
Supplier:  Sino Biological 11755H20B50
Encompass_Preferred
A DNA sequence encoding the human ADRBK1 (NP_001610.2) (Met 1-Leu 689) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7451