Learn More
Sino Biological Recombinant Human Semaphorin 4A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11756H08H100
Encompass_Preferred
A DNA sequence encoding the human SEMA4A (NP_071762.2) extracellular domain (Met 1-His 683) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7454