Learn More
Sino Biological Recombinant Human DPP10 Protein (His Tag)
Supplier:  Sino Biological 11760H07H100
Encompass_Preferred
A DNA sequence encoding the human DPP10 isoform 1 (AAH30832.1) extracellular domain (Leu56-Glu796) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7456