Learn More
Sino Biological Recombinant Human CALML5 Protein (His & GST Tag)
Supplier:  Sino Biological 11783H20E100
Encompass_Preferred
A DNA sequence encoding the human CALML5 (AAH39172.1) (Met 1-Glu 146) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7466