Learn More
Sino Biological Recombinant Human ACRV1 Protein (His Tag)
Supplier:  Sino Biological 11789H08H100
Encompass_Preferred
A DNA sequence encoding the mature form of human ACRV1 isoform 1 (P26436-1) (Gln 22-Ile 265) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-7468