Learn More
Sino Biological Recombinant Human M-CSF/CSF1 Protein (hFc Tag)
Supplier:  Sino Biological 11792H02H100
Encompass_Preferred
A DNA sequence encoding the human CSF1 (NP_757349.1) (Met1-Arg255) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7473