Learn More
Sino Biological Recombinant Human Siglec-5 Protein (hFc Tag)
Supplier:  Sino Biological 11798H02H100
Encompass_Preferred
A DNA sequence encoding the human SIGLEC5 (O15389) (Met1-Thr 434) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7486