Learn More
Sino Biological Recombinant Human Munc18c/STXBP3 Protein (His & GST Tag)
Supplier:  Sino Biological 11823H20B100
Encompass_Preferred
A DNA sequence encoding the human STXBP3 (O00186-1) (Met 1-Glu 592) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7502