Learn More
Sino Biological Recombinant Human Semaphorin 4D Protein (His Tag)
Supplier:  Sino Biological 11825H08H50
Encompass_Preferred
A DNA sequence encoding the human SEMA4D (AAH54500.1) extracellular domain (Met 1-Arg 734) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7507