Learn More
Sino Biological Recombinant Human SIRP gamma/SIRPG Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11828H08H100
Encompass_Preferred
A DNA sequence encoding the human SIRPG (NP_061026.2) extracellular domain (Met1-Ser364) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7510