Learn More
Sino Biological Recombinant Human PTK9 Protein (His & GST Tag)
Supplier:  Sino Biological 11829H20E100
Encompass_Preferred
A DNA sequence encoding the human TWF1 isoform 4 (Q12792-4) (Met 1-Asp 252) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7512