Learn More
Sino Biological Recombinant Human PTK9 Protein
Supplier:  Sino Biological 11829HNCE100
Encompass_Preferred
A DNA sequence encoding the human TWF1 isoform 4 (Q12792-4) (Met 1-Asp 252) was expressed and purified, with additional two amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-7514