Learn More
Sino Biological Recombinant Human CD300C/LMIR2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11832H08H100
Encompass_Preferred
A DNA sequence encoding the human CD300C (NP_006669.1) extracellular domain (Met 1-Arg 183) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7518