Learn More
Sino Biological Recombinant Human L-selectin/CD62L Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11838H08H100
Encompass_Preferred
A DNA sequence encoding the human SELL (P14151-1) extracellular domain (Met 1-Asn 332) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7524