Learn More
Sino Biological Recombinant Human CNTF Protein (His Tag)
Supplier:  Sino Biological 11841H07E20
Encompass_Preferred
A DNA sequence encoding the human CNTF (NP_000605.1) (Ala 2-Met 200) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7527