Learn More
Sino Biological Recombinant Human Neurexin-3 beta/NRXN3 Protein (His Tag)
Supplier:  Sino Biological 11843H08H100
Encompass_Preferred
A DNA sequence encoding the human NRXN3 beta isoform 2 (NP_620426.2) extracellular domain (Met 1-Thr 357) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7531