Learn More
Sino Biological Recombinant Human EDA2R/TNFRSF27 Protein (hFc Tag)
Supplier:  Sino Biological 11845H02H100
Encompass_Preferred
A DNA sequence encoding the human EDA2R (NP_068555.1) (Met1-Glu136) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7536