Learn More
Sino Biological Recombinant Human EDA2R/TNFRSF27 Protein (His Tag)
Supplier:  Sino Biological 11845H08H100
Encompass_Preferred
A DNA sequence encoding the human EDA2R (NP_068555.1) extracellular domain (Met 1-Thr 138) fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-7538