Learn More
Sino Biological Recombinant Human IL-4 Protein, HPLC-verified
Supplier:  Sino Biological 11846HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) was expressed and purified with an initial Met.
Catalog No. 50-161-7541