Learn More
Sino Biological Recombinant Human IL-17F Protein (His Tag)
Supplier:  Sino Biological 11855H07H100
Encompass_Preferred
A DNA sequence encoding the human IL17F (NP_443104.1) (Arg 31-Gln 163) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7547