Learn More
Sino Biological Recombinant Human IL-3 Protein (His Tag)
Supplier:  Sino Biological 11858H08H100
Encompass_Preferred
A DNA sequence encoding the human IL3 (NP_000579.2) (Met1-Phe152) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7561