Learn More
Sino Biological Recombinant Human HABP1/C1QBP/gC1qR Protein (His Tag)
Supplier:  Sino Biological 11874H08E100
Encompass_Preferred
A DNA sequence encoding the mature form of human C1QBP (NP_001203.1) (His 75-Gln 282) fused with two Met at N-terminus and a polyhistide tag at the C-terminus was expressed and purified.
Catalog No. 50-161-7573