Learn More
Sino Biological Recombinant Human IL-28B/Interferon lambda-3 Protein (His Tag)
Supplier:  Sino Biological 11890H08H50
Encompass_Preferred
A DNA sequence encoding the human IL28B (NP_742151.2) (Met 1-Val 196) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7582