Learn More
Sino Biological Recombinant Human IRE1 Protein (aa 465-977, His & GST Tag), HPLC-verified
Supplier:  Sino Biological 11905H20B50
Encompass_Preferred
A DNA sequence encoding the human ERN1 isoform 1 (O75460-1) cytoplasmic domain (Pro 465-Leu 977) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7593