Learn More
Sino Biological Recombinant Human MUSK Protein (aa 433-783, His & GST Tag)
Supplier:  Sino Biological 11918H20B50
Encompass_Preferred
A DNA sequence encoding the C-terminal segment of human MUSK isoform 2 (O15146-2) (Arg 433-Val 783) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7605