Learn More
Sino Biological Recombinant Human CPLX2 Protein (His Tag)
Supplier:  Sino Biological 11927H07E100
Encompass_Preferred
A DNA sequence encoding the human CPLX2 (Q6PUV4-1) (Asp 2-Lys 134) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7612