Learn More
Sino Biological Recombinant Human CAMKI Protein
Supplier:  Sino Biological 11932HNCE50
Encompass_Preferred
A DNA sequence encoding the human CAMK1 (NP_003647.1) (Leu 2-Leu 370) was expressed and purified, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-7619