Learn More
Sino Biological Recombinant Human TACI Protein (ECD, His Tag)
Supplier:  Sino Biological 11937H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF13B isoform 2 (O14836-2) extracellular domain (Ser 2-Thr 120) was expressed, with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-7628