Learn More
Sino Biological Recombinant Human IL17B Protein (hFc Tag)
Supplier:  Sino Biological 11939H02B100
Encompass_Preferred
A DNA sequence encoding the human IL17B (NP_055258.1) (Met1-Phe180) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7630