Learn More
Sino Biological Recombinant Human ANP32A Protein (His & GST Tag)
Supplier:  Sino Biological 11942H20E100
Encompass_Preferred
A DNA sequence encoding the of human ANP32A (NP_006296.1) (Glu 2-Lys 238) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7633