Learn More
Sino Biological Recombinant Human SLAMF6 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11945H08H100
Encompass_Preferred
A DNA sequence encoding the human SLAMF6 (Q96DU3-1) extracellular domain (Met 1-Met 226) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7637